Merck
  • Discovery of a Novel and Brain-Penetrant O-GlcNAcase Inhibitor via Virtual Screening, Structure-Based Analysis, and Rational Lead Optimization.
Discovery of a Novel and Brain-Penetrant O-GlcNAcase Inhibitor via Virtual Screening, Structure-Based Analysis, and Rational Lead Optimization.
Journal of medicinal chemistry (2021-01-07)
Michiko Tawada, Makoto Fushimi, Kei Masuda, Huikai Sun, Noriko Uchiyama, Yohei Kosugi, Weston Lane, Richard Tjhen, Satoshi Endo, Tatsuki Koike
ABSTRACT
O-GlcNAcase (OGA) has received increasing attention as an attractive therapeutic target for tau-mediated neurodegenerative disorders; however, its role in these pathologies remains unclear. Therefore, potent chemical tools with favorable pharmacokinetic profiles are desirable to characterize this enzyme. Herein, we report the discovery of a potent and novel OGA inhibitor, compound 5i, comprising an aminopyrimidine scaffold, identified by virtual screening based on multiple methodologies combining structure-based and ligand-based approaches, followed by sequential optimization with a focus on ligand lipophilicity efficiency. This compound was observed to increase the level of O-GlcNAcylated protein in cells and display suitable pharmacokinetic properties and brain permeability. Crystallographic analysis revealed that the chemical series bind to OGA via characteristic hydrophobic interactions, which resulted in a high affinity for OGA with moderate lipophilicity. Compound 5i could serve as a useful chemical probe to help establish a proof-of-concept of OGA inhibition as a therapeutic target for the treatment of tauopathies.
Sigma-Aldrich
TP-040n, ≥98% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
TP-040, ≥98% (HPLC)
Pricing and availability is not currently available.