Merck
  • Substance P/neurokinin-1 receptor pathway blockade ameliorates limbal stem cell deficiency by modulating mTOR pathway and preventing cell senescence.
Substance P/neurokinin-1 receptor pathway blockade ameliorates limbal stem cell deficiency by modulating mTOR pathway and preventing cell senescence.
Stem cell reports (2022-03-26)
Romina Lasagni Vitar, Francesca Triani, Marco Barbariga, Philippe Fonteyne, Paolo Rama, Giulio Ferrari
ABSTRACT
Severe ocular surface diseases can lead to limbal stem cell deficiency (LSCD), which is accompanied by defective healing. We aimed to evaluate the role of the substance P (SP)/neurokinin-1 receptor (NK1R) pathway in corneal epithelium wound healing in a pre-clinical model of LSCD. SP ablation or NK1R blockade significantly increased epithelial wound healing (p < 0.001) and corneal transparency (p < 0.001), compared with wild type (WT). In addition, a reduced number of infiltrating goblet and conjunctival cells (p < 0.05) and increased number of epithelial stem cells (p < 0.01), which also expressed NK1R, was observed. The mammalian target of rapamycin (mTOR) pathway was significantly inhibited (p < 0.05) and expression of γH2AX was significantly reduced (p < 0.05) after SP ablation. These results suggest that excessive expression of SP is associated with LSCD and results in accelerated senescence and exhaustion of residual stem cells. Topical treatment with NK1R antagonist ameliorates clinical signs associated with LSCD and could be used as an adjuvant treatment in LSCD.
Sigma-Aldrich
Anti-β-Actin−Peroxidase antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Pricing and availability is not currently available.