Merck
  • Genetic basis and adaptation trajectory of soybean from its temperate origin to tropics.
Genetic basis and adaptation trajectory of soybean from its temperate origin to tropics.
Nature communications (2021-09-16)
Lidong Dong, Chao Fang, Qun Cheng, Tong Su, Kun Kou, Lingping Kong, Chunbao Zhang, Haiyang Li, Zhihong Hou, Yuhang Zhang, Liyu Chen, Lin Yue, Lingshuang Wang, Kai Wang, Yongli Li, Zhuoran Gan, Xiaohui Yuan, James L Weller, Sijia Lu, Fanjiang Kong, Baohui Liu
ABSTRACT
Soybean (Glycine max) serves as a major source of protein and edible oils worldwide. The genetic and genomic bases of the adaptation of soybean to tropical regions remain largely unclear. Here, we identify the novel locus Time of Flowering 16 (Tof16), which confers delay flowering and improve yield at low latitudes and determines that it harbors the soybean homolog of LATE ELONGATED HYPOCOTYL (LHY). Tof16 and the previously identified J locus genetically additively but independently control yield under short-day conditions. More than 80% accessions in low latitude harbor the mutations of tof16 and j, which suggests that loss of functions of Tof16 and J are the major genetic basis of soybean adaptation into tropics. We suggest that maturity and yield traits can be quantitatively improved by modulating the genetic complexity of various alleles of the LHY homologs, J and E1. Our findings uncover the adaptation trajectory of soybean from its temperate origin to the tropics.
Sigma-Aldrich
Protein G Agarose/Salmon Sperm DNA, 2.5 mL, for use in chromatin immunoprecipitations (ChIP assays)