Merck
  • Chemical genetic screen identifies Gapex-5/GAPVD1 and STBD1 as novel AMPK substrates.
Chemical genetic screen identifies Gapex-5/GAPVD1 and STBD1 as novel AMPK substrates.
Cellular signalling (2019-02-18)
Serge Ducommun, Maria Deak, Anja Zeigerer, Olga Göransson, Susanne Seitz, Caterina Collodet, Agnete B Madsen, Thomas E Jensen, Benoit Viollet, Marc Foretz, Philipp Gut, David Sumpton, Kei Sakamoto
ABSTRACT
AMP-activated protein kinase (AMPK) is a key regulator of cellular energy homeostasis, acting as a sensor of energy and nutrient status. As such, AMPK is considered a promising drug target for treatment of medical conditions particularly associated with metabolic dysfunctions. To better understand the downstream effectors and physiological consequences of AMPK activation, we have employed a chemical genetic screen in mouse primary hepatocytes in an attempt to identify novel AMPK targets. Treatment of hepatocytes with a potent and specific AMPK activator 991 resulted in identification of 65 proteins phosphorylated upon AMPK activation, which are involved in a variety of cellular processes such as lipid/glycogen metabolism, vesicle trafficking, and cytoskeleton organisation. Further characterisation and validation using mass spectrometry followed by immunoblotting analysis with phosphorylation site-specific antibodies identified AMPK-dependent phosphorylation of Gapex-5 (also known as GTPase-activating protein and VPS9 domain-containing protein 1 (GAPVD1)) on Ser902 in hepatocytes and starch-binding domain 1 (STBD1) on Ser175 in multiple cells/tissues. As new promising roles of AMPK as a key metabolic regulator continue to emerge, the substrates we identified could provide new mechanistic and therapeutic insights into AMPK-activating drugs in the liver.
Sigma-Aldrich
Anti-GAPVD1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
Millipore
ANTI-FLAG® M2 Affinity Gel, purified immunoglobulin, buffered aqueous glycerol solution