Merck
  • Matrix Effects and Interferences of Different Citrus Fruit Coextractives in Pesticide Residue Analysis Using Ultrahigh-Performance Liquid Chromatography-High-Resolution Mass Spectrometry.
Matrix Effects and Interferences of Different Citrus Fruit Coextractives in Pesticide Residue Analysis Using Ultrahigh-Performance Liquid Chromatography-High-Resolution Mass Spectrometry.
Journal of agricultural and food chemistry (2017-05-26)
Natalia Besil, Verónica Cesio, Horacio Heinzen, Amadeo R Fernandez-Alba
ABSTRACT
The matrix effects of ethyl acetate extracts from seven different citrus fruits on the determination of 80 pesticide residues using liquid chromatography coupled to high-resolution time-of-flight mass spectrometry (UHPLC-(ESI)-HR-TOF) at 4 GHz resolution mode were studied. Only 20% of the evaluated pesticides showed noticeable matrix effects (ME) due to coelution with natural products between tR = 3 and 11 min. Principal component analysis (PCA) of the detected coextractives grouped the mandarins and the orange varieties, but separated lemon, oranges, and mandarins from each other. Matrix effects were different among species but similar between varieties, forcing the determination of pesticide residues through matrix-matched calibration curves with the same fruit. Twenty-three natural products (synephrine, naringin, poncirin, glycosides of hesperitin, limonin, nomilin, and a few fatty acids, among others) were identified in the analyzed extracts. Twelve of the identified compounds coeluted with 28 of the pesticides under study, causing different matrix effects.