Merck
  • A new strategy for the combination of supramolecular liquid phase microextraction and UV-Vis spectrophotometric determination for traces of maneb in food and water samples.
A new strategy for the combination of supramolecular liquid phase microextraction and UV-Vis spectrophotometric determination for traces of maneb in food and water samples.
Food chemistry (2020-09-20)
Mustafa Soylak, Metin Agirbas, Erkan Yilmaz
ABSTRACT
A novel and green method was developed for enrichment of maneb (manganese ethylene-bisdithiocarbamate) with a supramolecular solvent liquid phase microextraction method. The microextraction method has been used for the first time in the literature for separation-preconcentration of maneb. 1-decanol and tetrahydrofuran were used in the supramolecular solvent formation. The Mn2+ content of maneb was extracted in the supramolecular solvent phase as 1-(2-pyridylazo)-2-naphthol complex at pH 12.0. Manganese concentration was determined by UV-Vis spectrophotometer at 555 nm. Then, the maneb concentration equivalent to manganese concentration was calculated. The analytical parameters which effective in the method, including pH, volume of reagents, and sample volume were optimized. The limit of detection and the limit of quantification values for maneb were calculated as 2.22 μg L-1 and 7.32 μg L-1, respectively. The method was successfully applied in the analysis of the maneb content of water and food samples.
Product Description
Supelco
Maneb, PESTANAL®, analytical standard
Pricing and availability is not currently available.