Merck
  • Anti-neuroinflammatory capacity of fresh ginger is attributed mainly to 10-gingerol.
Anti-neuroinflammatory capacity of fresh ginger is attributed mainly to 10-gingerol.
Food chemistry (2013-07-23)
Su-Chen Ho, Ku-Shang Chang, Chih-Cheng Lin
ABSTRACT
Despite the anti-neuroinflammatory capacity of ginger, the corresponding active constituents are unclear. This study analyzed the composition of fresh ginger ethanolic extract by using LC-MS. Inhibitory activities of fresh ginger extract and seven gingerol-related compounds on the neuro-inflammation were also evaluated by using a lipopolysaccharide (LPS)-activated BV2 microglia culture model. Except for zingerone and 6-gingerol, other gingerols and shogaols at a concentration of 20 μM inhibited the production of nitric oxide, IL-1β, IL-6 and TNF-α as well as their mRNA levels in LPS-activated BV2 microglia. Blocking NF-κB activation was the underlying mechanism responsible for inhibiting the proinflammatory gene expression. Increasing the alkyl chain length enhanced the anti-neuroinflammatory capacity of gingerols yet, conversely, attenuated those of shogaols. 6-Gingerol was the most abundant compound in the fresh ginger extract, followed by 10-gingerol. Furthermore, fresh ginger extract exhibited a significant anti-neuroinflammatory capacity, which was largely owing to 10-gingerol, but not 6-gingerol.
Sigma-Aldrich
[10]-Gingerol, ≥98% (HPLC)