Merck
  • Very-low-density lipoprotein: complex particles in cardiac energy metabolism.
Very-low-density lipoprotein: complex particles in cardiac energy metabolism.
Journal of lipids (2011-07-21)
You-Guo Niu, Rhys D Evans
ABSTRACT
The heart is a major consumer of energy and is able to utilise a wide range of substrates including lipids. Nonesterified fatty acids (NEFA) were thought to be a favoured carbon source, but their quantitative contribution is limited because of their relative histotoxicity. Circulating triacylglycerols (TAGs) in the form of chylomicrons (CMs) and very-low-density lipoprotein (VLDL) are an alternative source of fatty acids and are now believed to be important in cardiac metabolism. However, few studies on cardiac utilisation of VLDL have been performed and the role of VLDL in cardiac energy metabolism remains unclear. Hearts utilise VLDL to generate ATP, but the oxidation rate of VLDL-TAG is relatively low under physiological conditions; however, in certain pathological states switching of energy substrates occurs and VLDL may become a major energy source for hearts. We review research regarding myocardial utilisation of VLDL and suggest possible roles of VLDL in cardiac energy metabolism: metabolic regulator and extracardiac energy storage for hearts.
Sigma-Aldrich
Lipoprotein, very low density from human plasma, ≥95% (SDS-PAGE), solution