- Synthesis of the functionalizable methionine surrogate azidohomoalanine using Boc-homoserine as precursor.
Synthesis of the functionalizable methionine surrogate azidohomoalanine using Boc-homoserine as precursor.
Nature protocols (2007-08-19)
A James Link, Mandy K S Vink, David A Tirrell
PMID17703199
ABSTRACT
This protocol describes a synthetic route to the non-canonical amino acid azidohomoalanine (AHA) using protected homoserine as a starting material. An alternative route to AHA is presented in a companion paper. This synthesis can be completed in 5 days.
MATERIALS