Merck
The Effects of Food on Cannabidiol Bioaccessibility.
Molecules (Basel, Switzerland) (2021-07-03)
Khadijeh Mozaffari, Stephanie Willette, Ben F Lucker, Sarah E Kovar, Francisco Omar Holguin, Ivette Guzman
ABSTRACT
Cannabidiol (CBD) is a hydrophobic non-psychoactive compound with therapeutic characteristics. Animal and human studies have shown its poor oral bioavailability in vivo, and the impact of consuming lipid-soluble CBD with and without food on gut bioaccessibility has not been explored. The purpose of this research was to study the bioaccessibility of CBD after a three-phase upper digestion experiment with and without food, and to test lipase activity with different substrate concentrations. Our results showed that lipase enzyme activity and fatty acid absorption increased in the presence of bile salts, which may also contribute to an increase in CBD bioaccessibility. The food matrix used was a mixture of olive oil and baby food. Overall, the fed-state digestion revealed significantly higher micellarization efficiency for CBD (14.15 ± 0.6% for 10 mg and 22.67 ± 2.1% for 100 mg CBD ingested) than the fasted state digestion of CBD (0.65 ± 0.7% for 10 mg and 0.14 ± 0.1% for 100 mg CBD ingested). The increase in bioaccessibility of CBD with food could be explained by the fact that micelle formation from hydrolyzed lipids aid in bioaccessibility of hydrophobic molecules. In conclusion, the bioaccessibility of CBD depends on the food matrix and the presence of lipase and bile salts.
Sigma-Aldrich
4-Nitrophenyl palmitate, lipase substrate