- Immunoaffinity purification of FLAG epitope-tagged bacterial alkaline phosphatase using a novel monoclonal antibody and peptide elution.
Immunoaffinity purification of FLAG epitope-tagged bacterial alkaline phosphatase using a novel monoclonal antibody and peptide elution.
BioTechniques (1994-04-01)
B L Brizzard, R G Chubet, D L Vizard
PMID8024796
ABSTRACT
The FLAG epitope is an eight amino acid peptide (AspTyrLysAspAspAspAspLys) that is useful for immunoaffinity purification of fusion proteins. A monoclonal antibody (anti-FLAG M1) that binds the FLAG epitope in a calcium-dependent manner and requires an N-terminal FLAG sequence has been described previously. We describe the use of a second anti-FLAG monoclonal antibody (anti-FLAG M2) in immunoaffinity purification of N-terminal Met-FLAG and C-terminal FLAG fusion to bacterial alkaline phosphatase. Although binding of an anti-FLAG M2 monoclonal antibody to the FLAG epitope is not calcium-dependent, bound fusion proteins can be eluted by competition with FLAG peptide.
Product Description
Millipore
ANTI-FLAG® M2 Affinity Gel, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Ubiquitin human, ≥95% (SDS-PAGE), recombinant, expressed in E. coli (N-terminal FLAG® tagged), lyophilized powder
Millipore
ANTI-FLAG TAG (DYKDDDDK) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution