Merck
  • The Effects of Sperm and Seminal Fluid of Immunized Male Mice on In Vitro Fertilization and Surrogate Mother-Embryo Interaction.
The Effects of Sperm and Seminal Fluid of Immunized Male Mice on In Vitro Fertilization and Surrogate Mother-Embryo Interaction.
International journal of molecular sciences (2021-10-14)
Galina Vladimirovna Kontsevaya, Ludmila Alekseevna Gerlinskaya, Yury Mikhailovich Moshkin, Margarita Vladimirovna Anisimova, Aliya Konstantinovna Stanova, Tatyana Ivanovna Babochkina, Mikhail Pavlovich Moshkin
ABSTRACT
The latest vaccination campaign has actualized the potential impact of antigenic stimuli on reproductive functions. To address this, we mimicked vaccination's effects by administering keyhole limpet hemocyanin (KLH ) to CD1 male mice and used their sperm for in vitro fertilization (IVF). Two-cell embryos after IVF with spermatozoa from control (C) or KLH-treated (Im) male mice were transferred to surrogate mothers mated with vasectomized control (C) or KLH-treated (Im) male mice, resulting in four experimental groups: C-C, Im-C, C-Im, and Im-Im. The pre-implantation losses were significantly lower in the Im-C group than in the C-Im group. At the same time, the resorption rates reduced markedly in the C-Im compared to the Im-C group. Embryo and placenta weights were significantly higher in the Im-Im group. Although the GM-CSF levels were lower in the amniotic fluid of the gestating surrogate mothers in the Im-Im group, they were strongly correlated with embryo mass. The number-size trade-off was only significant in the Im-Im group. This suggests a positive, cooperative effect of spermatozoa and seminal fluid from immune-primed males on embryo growth and the optimal distribution of surrogate mother maternal resources despite the negative impact of males' antigenic challenge on the IVF success rate.
Sigma-Aldrich
Hemocyanin-Keyhole Limpet, Native