Merck
  • Cyr61 promotes Schwann cell proliferation and migration via αvβ3 integrin.
Cyr61 promotes Schwann cell proliferation and migration via αvβ3 integrin.
BMC molecular and cell biology (2021-04-09)
Zhenghui Cheng, Yawen Zhang, Yinchao Tian, Yuhan Chen, Fei Ding, Han Wu, Yuhua Ji, Mi Shen
ABSTRACT
Schwann cells (SCs) play a crucial role in the repair of peripheral nerves. This is due to their ability to proliferate, migrate, and provide trophic support to axon regrowth. During peripheral nerve injury, SCs de-differentiate and reprogram to gain the ability to repair nerves. Cysteine-rich 61 (Cyr61/CCN1) is a member of the CCN family of matrix cell proteins and have been reported to be abundant in the secretome of repair mediating SCs. In this study we investigate the function of Cyr61 in SCs. We observed Cyr61 was expressed both in vivo and in vitro. The promoting effect of Cyr61 on SC proliferation and migration was through autocrine and paracrine mechanisms. SCs expressed αvβ3 integrin and the effect of Cyr61 on SC proliferation and migration could be blocked via αvβ3 integrin. Cyr61 could influence c-Jun protein expression in cultured SCs. In this study, we found that Cyr61 promotes SC proliferation and migration via αvβ3 integrin and regulates c-Jun expression. Our study contributes to the understanding of cellular and molecular mechanisms underlying SC's function during nerve injury, and thus, may facilitate the regeneration of peripheral nerves after injury.
Sigma-Aldrich
Monoclonal Anti-Thy 1.1 antibody produced in mouse, clone TN-26, purified immunoglobulin, buffered aqueous solution
Pricing and availability is not currently available.