Merck
  • Oxidative Aminoarylselenation of Maleimides via Copper-Catalyzed Four-Component Cross-Coupling.
Oxidative Aminoarylselenation of Maleimides via Copper-Catalyzed Four-Component Cross-Coupling.
Organic letters (2019-01-15)
Xue Gao, Liyang Tang, Lehao Huang, Zu-Sheng Huang, Yunfei Ma, Ge Wu
ABSTRACT
The first example of copper-catalyzed four-component coupling reaction of aryl iodides, Se powder, secondary amines, and maleimides is developed. This reaction provides an efficient and concise route to access aminoarylselenated maleimides via double C-Se bonds and C-N bond formation. The appealing features of this transformation are the use of Se powder as a selenating reagent, a green catalytic system, a wide range of substrate scope, and late-stage selenation of bioactive compounds.
Sigma-Aldrich
N-Methylpentylamine, 98%