Merck
  • Biochemical, genetic and physiological characterization of venom components from two species of scorpions: Centruroides exilicauda Wood and Centruroides sculpturatus Ewing.
Biochemical, genetic and physiological characterization of venom components from two species of scorpions: Centruroides exilicauda Wood and Centruroides sculpturatus Ewing.
Biochimie (2004-09-11)
Norma A Valdez-Cruz, Sonia Dávila, Alexei Licea, Miguel Corona, Fernando Z Zamudio, Jesús García-Valdes, Leslie Boyer, Lourival D Possani
ABSTRACT
Current literature concerning the taxonomic names of two possibly distinct species of scorpions from the genus Centruroides (sculpturatus and/or exilicauda) is controversial. This communication reports the results of biochemical, genetic and electrophysiological experiments conducted with C. exilicauda Wood of Baja California (Mexico) and C. sculpturatus Ewing of Arizona (USA). The chromatographic profile fractionation of the soluble venom from both species of scorpions is different. The N-terminal amino acid sequence for nine toxins of C. exilicauda was determined and compared with those from C. sculpturatus. Lethality tests conducted in mice support the idea that C. exilicauda venom should be expected to be medically less important than C. sculpturatus. Thirteen genes from the venomous glands of the scorpion C. exilicauda were obtained and compared with previously published sequences from genes of the species C. sculpturatus. Genes coding for cytochrome oxidase I and II of both species were also sequenced. A phylogenetic tree was generated with this information showing important differences between them. Additionally, the results of electrophysiological assays conducted with the venom from both species on the Ca(2+)-dependent K(+)-channels, showed significant differences. These results strongly support the conclusion that C. exilicauda and C. sculpturatus are in fact two distinct species of scorpions.