Merck
  • Novel insights into renal D-amino acid oxidase accumulation: propiverine changes DAAO localization and peroxisomal size in vivo.
Novel insights into renal D-amino acid oxidase accumulation: propiverine changes DAAO localization and peroxisomal size in vivo.
Archives of toxicology (2016-03-11)
Lisanne Luks, Silvia Sacchi, Loredano Pollegioni, Daniel R Dietrich
ABSTRACT
Chronic exposure to propiverine, a frequently prescribed pharmaceutical for treatment of overactive bladder and incontinence, provokes massive protein accumulation in the cytosol and nucleus of renal proximal tubule epithelial cells in rats. Previously, the accumulating protein was identified as D-amino acid oxidase (DAAO), a peroxisomal flavoenzyme expressed in kidney, liver and brain. The cellular mechanism of propiverine-induced DAAO accumulation, however, remains unexplained and poorly characterized. Therefore, to further increase the understanding of DAAO accumulation in rat kidney, this study aimed to characterize DAAO accumulations using differential immunofluorescent staining of rat kidney sections as well as in vitro binding analyses and proteasomal activity studies. We demonstrated that propiverine is neither a ligand of DAAO nor an inhibitor of the proteasome in vitro. However, propiverine treatment resulted in a significant decrease of peroxisomal size in rat proximal tubule epithelial cells. Moreover, peroxisomal catalase also accumulated in the cytosol and nuclei of propiverine-treated rats concurrently with DAAO. Taken together, our study indicates that propiverine treatment affects the trafficking and/or degradation of peroxisomal proteins such as DAAO and catalase by a so far unique and unknown mechanism.