Merck
  • DUSP11-mediated control of 5'-triphosphate RNA regulates RIG-I sensitivity.
DUSP11-mediated control of 5'-triphosphate RNA regulates RIG-I sensitivity.
Genes & development (2020-11-14)
Joon H Choi, James M Burke, Kayla H Szymanik, Upasana Nepal, Anna Battenhouse, Justin T Lau, Aaron Stark, Victor Lam, Christopher S Sullivan
ABSTRACT
Deciphering the mechanisms that regulate the sensitivity of pathogen recognition receptors is imperative to understanding infection and inflammation. Here we demonstrate that the RNA triphosphatase dual-specificity phosphatase 11 (DUSP11) acts on both host and virus-derived 5'-triphosphate RNAs rendering them less active in inducing a RIG-I-mediated immune response. Reducing DUSP11 levels alters host triphosphate RNA packaged in extracellular vesicles and induces enhanced RIG-I activation in cells exposed to extracellular vesicles. Virus infection of cells lacking DUSP11 results in a higher proportion of triphosphorylated viral transcripts and attenuated virus replication, which is rescued by reducing RIG-I expression. Consistent with the activity of DUSP11 in the cellular RIG-I response, mice lacking DUSP11 display lower viral loads, greater sensitivity to triphosphorylated RNA, and a signature of enhanced interferon activity in select tissues. Our results reveal the importance of controlling 5'-triphosphate RNA levels to prevent aberrant RIG-I signaling and demonstrate DUSP11 as a key effector of this mechanism.
Sigma-Aldrich
MISSION® esiRNA, targeting human DDX58