Merck
  • Ultra-stretchable, self-recovering, self-healing cationic guar gum/poly(stearyl methacrylate-co-acrylic acid) hydrogels.
Ultra-stretchable, self-recovering, self-healing cationic guar gum/poly(stearyl methacrylate-co-acrylic acid) hydrogels.
Carbohydrate polymers (2021-01-24)
Houchao Jing, Jinyang Feng, Jianping Shi, Lin He, Peipei Guo, Shuang Guan, Hai Fu, Yuhui Ao
ABSTRACT
Hydrogels that exhibit properties such as ultra-elongation, self-recovery, and self-healing have applications in sensors and many other fields. With these properties and applications in mind, we hypothesised that we could develop a strain-sensing hydrogel based on acrylic acid, stearyl methacrylate, cationic guar gum, and hexadecyl trimethyl ammonium bromide, without any covalent crosslinker. The hydrogels are instead held together by physical, non-covalent interactions such as ionic interactions, hydrogen bonding, and the hydrophobic effect, as suggested by spectroscopy and swelling experiments. The hydrogels exhibit many useful properties, such as: excellent stretching-up to 4267%-and almost complete reversion to their original state at a large strain of 500%, even after 20 successive cycles; temperature-dependent self-healing and self-recovery; and strain-sensitive conductivity that is attributable to the directional migration of ions. Because of these outstanding features, such as notch-insensitivity and the ability to withstand knotting under high strain, our hydrogels will be useful as flexible sensors.
Sigma-Aldrich
Solketal methacrylate, 50 wt. % in dichloromethane, contains ~280 ppm 4-tert-butylcatechol as inhibitor
Sigma-Aldrich
Stearyl methacrylate, Mixture of stearyl and cetyl methacrylates, contains MEHQ as inhibitor