Merck
  • A stable cathode-solid electrolyte composite for high-voltage, long-cycle-life solid-state sodium-ion batteries.
A stable cathode-solid electrolyte composite for high-voltage, long-cycle-life solid-state sodium-ion batteries.
Nature communications (2021-02-25)
Erik A Wu, Swastika Banerjee, Hanmei Tang, Peter M Richardson, Jean-Marie Doux, Ji Qi, Zhuoying Zhu, Antonin Grenier, Yixuan Li, Enyue Zhao, Grayson Deysher, Elias Sebti, Han Nguyen, Ryan Stephens, Guy Verbist, Karena W Chapman, Raphaële J Clément, Abhik Banerjee, Ying Shirley Meng, Shyue Ping Ong
ABSTRACT
Rechargeable solid-state sodium-ion batteries (SSSBs) hold great promise for safer and more energy-dense energy storage. However, the poor electrochemical stability between current sulfide-based solid electrolytes and high-voltage oxide cathodes has limited their long-term cycling performance and practicality. Here, we report the discovery of the ion conductor Na3-xY1-xZrxCl6 (NYZC) that is both electrochemically stable (up to 3.8 V vs. Na/Na+) and chemically compatible with oxide cathodes. Its high ionic conductivity of 6.6 × 10-5 S cm-1 at ambient temperature, several orders of magnitude higher than oxide coatings, is attributed to abundant Na vacancies and cooperative MCl6 rotation, resulting in an extremely low interfacial impedance. A SSSB comprising a NaCrO2 + NYZC composite cathode, Na3PS4 electrolyte, and Na-Sn anode exhibits an exceptional first-cycle Coulombic efficiency of 97.1% at room temperature and can cycle over 1000 cycles with 89.3% capacity retention at 40 °C. These findings highlight the immense potential of halides for SSSB applications.
Sigma-Aldrich
Zirconium(IV) chloride, anhydrous, powder, 99.99% trace metals basis
Pricing and availability is not currently available.