Merck
  • In-situ visualization of the space-charge-layer effect on interfacial lithium-ion transport in all-solid-state batteries.
In-situ visualization of the space-charge-layer effect on interfacial lithium-ion transport in all-solid-state batteries.
Nature communications (2020-11-20)
Longlong Wang, Ruicong Xie, Bingbing Chen, Xinrun Yu, Jun Ma, Chao Li, Zhiwei Hu, Xingwei Sun, Chengjun Xu, Shanmu Dong, Ting-Shan Chan, Jun Luo, Guanglei Cui, Liquan Chen
ABSTRACT
The space charge layer (SCL) is generally considered one of the origins of the sluggish interfacial lithium-ion transport in all-solid-state lithium-ion batteries (ASSLIBs). However, in-situ visualization of the SCL effect on the interfacial lithium-ion transport in sulfide-based ASSLIBs is still a great challenge. Here, we directly observe the electrode/electrolyte interface lithium-ion accumulation resulting from the SCL by investigating the net-charge-density distribution across the high-voltage LiCoO2/argyrodite Li6PS5Cl interface using the in-situ differential phase contrast scanning transmission electron microscopy (DPC-STEM) technique. Moreover, we further demonstrate a built-in electric field and chemical potential coupling strategy to reduce the SCL formation and boost lithium-ion transport across the electrode/electrolyte interface by the in-situ DPC-STEM technique and finite element method simulations. Our findings will strikingly advance the fundamental scientific understanding of the SCL mechanism in ASSLIBs and shed light on rational electrode/electrolyte interface design for high-rate performance ASSLIBs.
Sigma-Aldrich
2-Methoxyethanol, anhydrous, 99.8%
Pricing and availability is not currently available.