Merck
  • The use of model cellulose gel beads to clarify flame-retardant characteristics of layer-by-layer nanocoatings.
The use of model cellulose gel beads to clarify flame-retardant characteristics of layer-by-layer nanocoatings.
Carbohydrate polymers (2021-01-14)
Oruç Köklükaya, Rose-Marie Pernilla Karlsson, Federico Carosio, Lars Wågberg
ABSTRACT
Layer-by-Layer (LbL) assembled nanocoatings are exploited to impart flame-retardant properties to cellulosic substrates. A model cellulose material can make it possible to investigate an optimal bilayer (BL) range for the deposition of coating while elucidating the main flame-retardant action thus allowing for an efficient design of optimized LbL formulations. Model cellulose gel beads were prepared by dissolving cellulose-rich fibers followed by precipitation. The beads were LbL-treated with chitosan (CH) and sodium hexametaphosphate (SHMP). The char forming properties were studied using thermal gravimetric analysis. The coating increased the char yield in nitrogen to up to 29 % and showed a distinct pattern of micro intumescent behavior upon heating. An optimal range of 10-20 BL is observed. The well-defined model cellulose gel beads hence introduce a new scientific route both to clarify the fundamental effects of different film components and to optimize the composition of the films.
Sigma-Aldrich
Ethanol – 2-propanol mixture, BioUltra, for molecular biology, 19:1, A15 ISO1, ≥99.5% (GC)