Merck
  • Bile salt-dependent lipase promotes the barrier integrity of Caco-2 cells by activating Wnt/β-catenin signaling via LRP6 receptor.
Bile salt-dependent lipase promotes the barrier integrity of Caco-2 cells by activating Wnt/β-catenin signaling via LRP6 receptor.
Cell and tissue research (2020-11-28)
Yaqi Qiu, Jiefei Zhou, Dandan Zhang, Huanlei Song, Linxi Qian
ABSTRACT
Bile salt-dependent lipase (BSDL) within intestinal lumen can be endocytosed by enterocytes and support the intestinal barrier function. However, the epithelial-supporting effect of this protein has not been verified in a human cell line and neither the direct signaling pathway nor the function of endocytosis in this process has been clearly identified. We sought to investigate the signaling pathway and the membrane receptor through which BSDL might exert these effects using intestinal epithelial cells. Caco-2 cells were treated with recombinant BSDL, and the barrier function, cell proliferation, and activation of the Wnt signaling pathway were assessed. The effect of Wnt signaling activation induced by BSDL and BSDL endocytosis was investigated in LRP6-silenced and non-silenced cells. Moreover, caveolae- and clathrin-dependent endocytosis inhibitors were also applied respectively to analyze their effects on Wnt signaling activation induced by BSDL. BSDL treatment increased the barrier function but not proliferation of Caco-2 cells. It also induced β-catenin nuclear translocation and activated Wnt target gene transcription. Moreover, in the Wnt pathway, BSDL increased the levels of non-phosphorylated-β-catenin (Ser33/37/Thr41) and phosphorylated-β-catenin (Ser552). Notably, the silencing of LRP6 expression impaired BSDL endocytosis and decreased BSDL-induced β-catenin nuclear translocation. The inhibition of BSDL endocytosis induced by caveolae-mediated endocytosis inhibitor was stronger than that by clathrin-mediated endocytosis inhibitor, and the Wnt signaling activation associated with its endocytosis was also most likely caveolae-dependent. Our findings suggested that LRP6, a canonical Wnt pathway co-receptor, can mediate BSDL endocytosis and then activate Wnt signaling in Caco-2 cells.