Merck
  • Selective activation of beta3-adrenoceptors by octopamine: comparative studies in mammalian fat cells.
Selective activation of beta3-adrenoceptors by octopamine: comparative studies in mammalian fat cells.
Naunyn-Schmiedeberg's archives of pharmacology (1999-05-27)
C Carpéné, J Galitzky, E Fontana, C Atgié, M Lafontan, M Berlan
ABSTRACT
Numerous synthetic agonists selectively stimulate beta3-adrenoceptors (ARs). The endogenous catecholamines, noradrenaline and adrenaline, however, stimulate all the beta-AR subtypes, and no selective physiological agonist for beta3-ARs has been described so far. The aim of this study was to investigate whether any naturally occurring amine can stimulate selectively beta3-ARs. Since activation of lipolysis is a well-known beta-adrenergic function, the efficacy and potency of various biogenic amines were compared with those of noradrenaline, isoprenaline, and beta3-AR agonists 4-(-{[2-hydroxy-(3-chlorophenyl)ethyl]-amino} propyl)phenoxyacetate (BRL 37,344) and (R,R)-5-(2-{[2-(3-chlorophenyl )-2-hydroxyethyl]-amino} propyl)-1,3-benzo-dioxole-2,2-dicarboxylate (CL 316,243) by testing their lipolytic action in white fat cells. Five mammalian species were studied: rat, hamster and dog, in which selective beta-AR agonists act as full lipolytic agents, and guinea-pigs and humans, in which beta3-AR agonists are less potent activators of lipolysis. Several biogenic amines were inefficient (e.g. dopamine, tyramine and beta-phenylethylamine) while others (synephrine, phenylethanolamine, epinine) were partially active in stimulating lipolysis in all species studied. Their actions were inhibited by all the beta-AR antagonists tested, including those selective for beta1- or beta2-ARs. Octopamine was the only amine fully stimulating lipolysis in rat, hamster and dog fat cells, while inefficient in guinea-pig or human fat cells, like the beta3-AR agonists. In rat white fat cells, beta-AR antagonists inhibited the lipolytic effect of octopamine with a relative order of potency very similar to that observed against CL 316,243. Competitive antagonism of octopamine effect resulted in the following apparent pA2 [-log(IC50), where IC50 is the antagonist concentration eliciting half-maximal inhibition] values: 7.77 (bupranolol), 6.48 [3-(2-ethyl-phenoxy)-1[(1 S)-1,2,3,4-tetrahydronaphth-1-ylaminol]-(2S)2-propanol oxalate, SR 59230A, a beta3-selective antagonist], 6.30[erythro-D,L-1(7-lethylindan-4-yloxy)-3-isopropylamino-+ ++butan-2-ol, ICI 118,551, a beta2-selective antagonist] and 4.71 [(+/-)-[2-(3-carbomyl-4-hydroxyphenoxy)-ethylamino]-3-[4-(1- methyl-4-trifluoromethyl-2-imidazolyl)-phenoxy]2-propanolmethane sulphonate, CGP 20712A, a beta1-selective antagonist]. Octopamine had other properties in common with beta3-AR agonists: stimulation of oxygen consumption in rat brown fat cells and very low affinity in displacing [3H]CGP 12,177 binding to [beta1- or beta2-ARs in dog and rat adipocyte membranes. In Chinese hamster ovary (CHO) cells expressing human beta3-ARs, octopamine inhibited [125I]ICYP binding with only twofold less affinity than noradrenaline while it exhibited an affinity around 200-fold lower than noradrenaline in CHO cells expressing human beta1- or beta2-ARs. These data suggest that, among the biogenic amines metabolically related to catecholamines, octopamine can be considered as the most selective for beta3-ARs.
Sigma-Aldrich
(±)-Synephrine, ≥98%