Merck
  • Comparison of Regular, Pure Shift, and Fast 2D NMR Experiments for Determination of the Geographical Origin of Walnuts.
Comparison of Regular, Pure Shift, and Fast 2D NMR Experiments for Determination of the Geographical Origin of Walnuts.
Metabolites (2021-01-13)
Stephanie Watermann, Caroline Schmitt, Tobias Schneider, Thomas Hackl
ABSTRACT
1H NMR spectroscopy, in combination with chemometric methods, was used to analyze the methanol/acetonitrile (1:1) extract of walnut (Juglans Regia L.) regarding the geographical origin of 128 authentic samples from different countries (France, Germany, China) and harvest years (2016-2019). Due to the large number of different metabolites within the acetonitrile/methanol extract, the one-dimensional (1D) 1H NOESY (nuclear Overhauser effect spectroscopy) spectra suffer from strongly overlapping signals. The identification of specific metabolites and statistical analysis are complicated. The use of pure shift 1H NMR spectra such as PSYCHE (pure shift yielded by chirp excitation) or two-dimensional ASAP-HSQC (acceleration by sharing adjacent polarization-heteronuclear single quantum correlation) spectra for multivariate analysis to determine the geographical origin of foods may be a promising method. Different types of NMR spectra (1D 1H NOESY, PSYCHE, and ASAP-HSQC) were acquired for each of the 128 walnut samples and the results of the statistical analysis were compared. A support vector machine classifier was applied for differentiation of samples from Germany/China, France/Germany, and France/China. The models obtained by conduction of a repeated nested cross-validation showed accuracies from 58.9% (±1.3%) to 95.9% (±0.8%). The potential of the 1H-13C HSQC as a 2D NMR experiment for metabolomics studies was shown.
Sigma-Aldrich
Octamethylcyclotetrasiloxane, 98%
Pricing and availability is not currently available.