Merck
  • Discovery of tetrahydrotetramethylnaphthalene analogs as adult T-cell leukemia cell-selective proliferation inhibitors in a small chemical library constructed based on multi-template hypothesis.
Discovery of tetrahydrotetramethylnaphthalene analogs as adult T-cell leukemia cell-selective proliferation inhibitors in a small chemical library constructed based on multi-template hypothesis.
Bioorganic & medicinal chemistry (2009-05-16)
Masahiko Nakamura, Takayuki Hamasaki, Maiko Tokitou, Masanori Baba, Yuichi Hashimoto, Hiroshi Aoyama
ABSTRACT
Adult T cell leukemia (ATL), caused by infection of human T-lymphotropic virus type 1 (HTLV-1), has a poor prognosis and curative therapy is unavailable, so it is important to find or design superior lead compounds for the drug treatment of ATL. We used our micro-reversed fragment-based drug design hypothesis and multi-template hypothesis to extract the tetrahydrotetramethylnaphthalene (TMN) skeleton from tamibarotene, a useful medicament for the treatment of acute promyelocytic leukemia (APL). Structural development of TMN yielded highly ATL cell-selective growth inhibitors, including 2-acetyl-3-hydroxy-5,6,7,8-tetrahydro-5,5,8,8-tetramethylnaphthalene (6). Structure-activity relationship analysis suggests the existence of a specific target molecule for ATL cell-selective inhibition of proliferation through G2 arrest.
Sigma-Aldrich
2′,4′-Dimethylacetophenone, 96%