Merck
  • Cardiolipin exposure on the outer mitochondrial membrane modulates α-synuclein.
Cardiolipin exposure on the outer mitochondrial membrane modulates α-synuclein.
Nature communications (2018-02-28)
Tammy Ryan, Vladimir V Bamm, Morgan G Stykel, Carla L Coackley, Kayla M Humphries, Rhiannon Jamieson-Williams, Rajesh Ambasudhan, Dick D Mosser, Stuart A Lipton, George Harauz, Scott D Ryan
ABSTRACT
Neuronal loss in Parkinson's disease (PD) is associated with aberrant mitochondrial function and impaired proteostasis. Identifying the mechanisms that link these pathologies is critical to furthering our understanding of PD pathogenesis. Using human pluripotent stem cells (hPSCs) that allow comparison of cells expressing mutant SNCA (encoding α-synuclein (α-syn)) with isogenic controls, or SNCA-transgenic mice, we show that SNCA-mutant neurons display fragmented mitochondria and accumulate α-syn deposits that cluster to mitochondrial membranes in response to exposure of cardiolipin on the mitochondrial surface. Whereas exposed cardiolipin specifically binds to and facilitates refolding of α-syn fibrils, prolonged cardiolipin exposure in SNCA-mutants initiates recruitment of LC3 to the mitochondria and mitophagy. Moreover, we find that co-culture of SNCA-mutant neurons with their isogenic controls results in transmission of α-syn pathology coincident with mitochondrial pathology in control neurons. Transmission of pathology is effectively blocked using an anti-α-syn monoclonal antibody (mAb), consistent with cell-to-cell seeding of α-syn.