Merck
  • Antioxidant, Physicochemical, and Cellular Secretion of Glucagon-Like Peptide-1 Properties of Oat Bran Protein Hydrolysates.
Antioxidant, Physicochemical, and Cellular Secretion of Glucagon-Like Peptide-1 Properties of Oat Bran Protein Hydrolysates.
Antioxidants (Basel, Switzerland) (2020-07-02)
Mallory E Walters, William G Willmore, Apollinaire Tsopmo
ABSTRACT
The aim of this work was to determine the physicochemical and biological activities of hydrolyzed proteins from sonicated oat brans. In addition to the control bran sample, two types of pre-treatment procedures-namely, ultrasonic bath and probe-type sonication-were performed to extract proteins, followed by hydrolysis with various proteases. Physicochemical analyses showed that Flavourzyme-hydrolysates had greater amounts of aromatic amino acids, Papain-hydrolysates low surface charges (-0.78 to -1.32 mV) compared to the others (-3.67 to -9.17 mV), and Alcalase-hydrolysates a higher surface hydrophobicity. The hydrolysates had good radical scavenging activities but, as the ultrasonic pre-treatment of the brans showed, in certain cases there was a reduction in activities of up to 22% for ROO• and HO• and 15% for O2•- radicals. In anti-diabetic tests, the maximum inhibition of α-amylase was 31.8%, while that of dipeptidyl peptidase-4 was 53.6%. In addition, the secretion of glucagon-like peptide-1 in NCI-H716 cells was enhanced by 11.5% in the presence of hydrolysates.
Sigma-Aldrich
ECM Gel from Engelbreth-Holm-Swarm murine sarcoma, liquid, BioReagent, suitable for cell culture
Millipore
Glucagon Like Peptide-1 (Active) ELISA, This Glucagon Like Peptide-1 (Active) ELISA is used to measure & quantify Glucagon Like Peptide-1 levels in Metabolism & Endocrine research.