Merck
  • The effect of forage-types on the fatty acid profile, lipid and protein oxidation, and retail colour stability of muscles from White Dorper lambs.
The effect of forage-types on the fatty acid profile, lipid and protein oxidation, and retail colour stability of muscles from White Dorper lambs.
Meat science (2017-04-26)
Gerlane F De Brito, Benjamin W B Holman, Shawn R McGrath, Michael A Friend, Remy van de Ven, David L Hopkins
ABSTRACT
The aim of this study was to evaluate the effect of different forage-types on lamb meat quality parameters. White Dorper lambs that had grazed five forage-types, were slaughtered commercially. At 24h post-mortem, the m. longissimus lumborum (LL) was removed from one side, sliced into three equal sub-samples, vacuum packaged and assigned to ageing periods (5, 12 or 40days); the other side of LL was aged for 5days. The m. adductor femoris was used for fatty acid analysis. Lambs fed chicory+arrowleaf clover had the highest concentration of health claimable omega-3 fatty acids and the lowest omega-6:omega-3 fatty acid ratio. Forage-types with higher vitamin E content showed lower lipid oxidation levels independent of ageing period. Forage-type and ageing period did not influence the redness, yellowness, chroma or reflectance ratio (630nm÷580nm) of displayed meat. Chicory+arrowleaf clover gave the best results to improve the fatty acid content of lamb meat.
Sigma-Aldrich
Nitrite/Nitrate Assay Kit, colorimetric, used for detection of nitric oxide metabolites
Sigma-Aldrich
Protein Carbonyl Content Assay Kit, sufficient for 100 colorimetric tests