Merck
  • Complement Genetic Variants and FH Desialylation in S. pneumoniae-Haemolytic Uraemic Syndrome.
Complement Genetic Variants and FH Desialylation in S. pneumoniae-Haemolytic Uraemic Syndrome.
Frontiers in immunology (2021-03-30)
Irene Gómez Delgado, Fernando Corvillo, Pilar Nozal, Emilia Arjona, Álvaro Madrid, Marta Melgosa, Juan Bravo, Ágnes Szilágyi, Dorottya Csuka, Nóra Veszeli, Zoltán Prohászka, Pilar Sánchez-Corral
ABSTRACT
Haemolytic Uraemic Syndrome associated with Streptococcus pneumoniae infections (SP-HUS) is a clinically well-known entity that generally affects infants, and could have a worse prognosis than HUS associated to E. coli infections. It has been assumed that complement genetic variants associated with primary atypical HUS cases (aHUS) do not contribute to SP-HUS, which is solely attributed to the action of the pneumococcal neuraminidase on the host cellular surfaces. We previously identified complement pathogenic variants and risk polymorphisms in a few Hungarian SP-HUS patients, and have now extended these studies to a cohort of 13 Spanish SP-HUS patients. Five patients presented rare complement variants of unknown significance, but the frequency of the risk haplotypes in the CFH-CFHR3-CFHR1 region was similar to the observed in aHUS. Moreover, we observed desialylation of Factor H (FH) and the FH-Related proteins in plasma samples from 2 Spanish and 4 Hungarian SP-HUS patients. To analyze the functional relevance of this finding, we compared the ability of native and "in vitro" desialylated FH in: (a) binding to C3b-coated microtiter plates; (b) proteolysis of fluid-phase and surface-bound C3b by Factor I; (c) dissociation of surface bound-C3bBb convertase; (d) haemolytic assays on sheep erythrocytes. We found that desialylated FH had reduced capacity to control complement activation on sheep erythrocytes, suggesting a role for FH sialic acids on binding to cellular surfaces. We conclude that aHUS-risk variants in the CFH-CFHR3-CFHR1 region could also contribute to disease-predisposition to SP-HUS, and that transient desialylation of complement FH by the pneumococcal neuraminidase may have a role in disease pathogenesis.
Sigma-Aldrich
Neuraminidase Activity Assay Kit, sufficient for 100 colorimetric or fluorometric tests