- Complete Genome Sequence of Paenibacillus polymyxa CR1, a Plant Growth-Promoting Bacterium Isolated from the Corn Rhizosphere Exhibiting Potential for Biocontrol, Biomass Degradation, and Biofuel Production.
Complete Genome Sequence of Paenibacillus polymyxa CR1, a Plant Growth-Promoting Bacterium Isolated from the Corn Rhizosphere Exhibiting Potential for Biocontrol, Biomass Degradation, and Biofuel Production.
Genome announcements (2014-01-25)
Alexander W Eastman, Brian Weselowski, Naeem Nathoo, Ze-Chun Yuan
PMID24459277
ABSTRACT
Here we report the complete genome sequence of the bacterium Paenibacillus polymyxa CR1 (accession no. CP006941), which consists of one circular chromosome of 6,024,666 bp with 5,283 coding sequences (CDS), 87 tRNAs, and 12 rRNA operons. Data presented will allow for further insights into the mechanisms underpinning agriculturally and industrially relevant processes.