Merck
  • Does selective serotonin reuptake inhibitor (SSRI) fluoxetine affects mussel Mytilus galloprovincialis?
Does selective serotonin reuptake inhibitor (SSRI) fluoxetine affects mussel Mytilus galloprovincialis?
Environmental pollution (Barking, Essex : 1987) (2012-12-04)
Maria Gonzalez-Rey, Maria João Bebianno
ABSTRACT
Fluoxetine (FLX) the active pharmaceutical ingredient (API) in Prozac(®) is a widely prescribed psychoactive drug which ubiquitous occurrence in the aquatic environment is associated to a poor removal rate in waste-water treatment plant (WWTP) systems. This API acts as a selective serotonin reuptake inhibitor (SSRI) frequently reported to cause disrupting effects in non-target species. The objective of this study includes a multibiomarker response evaluation on mussel Mytilus galloprovincialis during two weeks exposure to 75 ng L(-1) FLX assessing antioxidant enzymes activities--superoxide dismutase (SOD), catalase (CAT) and glutathione-S-transferase (GST); lipid peroxidation (LPO), acetylcholinesterase (AChE) neurotoxic response and endocrine disruption through alkali-labile phosphates (ALP) indirect measurement of vitellogenin-like proteins. Results show transient tissue-specific enzymatic responses and damage affecting mostly mussel gills. However, the clear ALP levels inhibition throughout time in both sex-differentiated gonads gives evidence to FLX reinforced action as an endocrine disruptor rather than an oxidative or neurotoxic inducer.
Sigma-Aldrich
Xanthine Oxidase from bovine milk, Grade I, ammonium sulfate suspension, ≥0.4 units/mg protein
Pricing and availability is not currently available.
Sigma-Aldrich
R-(−)-Fluoxetine hydrochloride, >98% (HPLC), solid
Pricing and availability is not currently available.