Merck
  • The adenylyl cyclase inhibitor MDL-12,330A potentiates insulin secretion via blockade of voltage-dependent K(+) channels in pancreatic beta cells.
The adenylyl cyclase inhibitor MDL-12,330A potentiates insulin secretion via blockade of voltage-dependent K(+) channels in pancreatic beta cells.
PloS one (2013-11-10)
Xiaodong Li, Qing Guo, Jingying Gao, Jing Yang, Wan Zhang, Yueqin Liang, Dongmei Wu, Yunfeng Liu, Jianping Weng, Qingshan Li, Yi Zhang
ABSTRACT
Adenylyl cyclases (ACs) play important role in regulating pancreatic beta cell growth, survival and secretion through the synthesis of cyclic AMP (cAMP). MDL-12,330A and SQ 22536 are two AC inhibitors used widely to establish the role of ACs. The goal of this study was to examine the effects of MDL-12,330A and SQ 22536 on insulin secretion and underlying mechanisms. Patch-clamp recording, Ca(2+) fluorescence imaging and radioimmunoassay were used to measure outward K(+) currents, action potentials (APs), intracellular Ca(2+) ([Ca(2+)]i) and insulin secretion from rat pancreatic beta cells. MDL-12,330A (10 µmol/l) potentiated insulin secretion to 1.7 times of control in the presence of 8.3 mmol/l glucose, while SQ 22536 did not show significant effect on insulin secretion. MDL-12,330A prolonged AP durations (APDs) by inhibiting voltage-dependent K(+) (KV) channels, leading to an increase in [Ca(2+)]i levels. It appeared that these effects induced by MDL-12,330A did not result from AC inhibition, since SQ 22536 did not show such effects. Furthermore, inhibition of the downstream effectors of AC/cAMP signaling by PKA inhibitor H89 and Epac inhibitor ESI-09, did not affect KV channels and insulin secretion. The putative AC inhibitor MDL-12,330A enhances [Ca(2+)]i and insulin secretion via inhibition of KV channels rather than AC antagonism in beta cells, suggesting that the non-specific effects is needed to be considered for the right interpretation of the experimental results using this agent in the analyses of the role of AC in cell function.
Sigma-Aldrich
MDL-12,330A hydrochloride, ≥98% (HPLC), powder
Pricing and availability is not currently available.