Merck
  • Synthesis, biological activity, and molecular modeling of selective 5-HT(2C/2B) receptor antagonists.
Synthesis, biological activity, and molecular modeling of selective 5-HT(2C/2B) receptor antagonists.
Journal of medicinal chemistry (1996-12-06)
I T Forbes, S Dabbs, D M Duckworth, P Ham, G E Jones, F D King, D V Saunders, F E Blaney, C B Naylor, G S Baxter, T P Blackburn, G A Kennett, M D Wood
ABSTRACT
The synthesis and biological activity are reported for a series of analogues of the previously published indole urea 2 (SB-206553), designed to probe the 5-HT(2C) receptor binding site. Small molecule modeling studies have been used to define a region in space which is allowed at the 5-HT(2C) receptor but disallowed at the 5-HT(2A) receptor. In a complementary approach, docking of 2 into our model of the 5-HT(2C) receptor has allowed us to propose a novel primary binding interaction for this series of diaryl ureas, involving a potential double hydrogen-bonding interaction between the urea carbonyl oxygen of the ligand and two serine residues in the receptor. The difference of two valine residues in the 5-HT(2C) receptor for leucine residues in the 5-HT(2A) receptor is believed to account for the observed 5-HT(2C)/5-HT(2A) selectivity with 2.
Sigma-Aldrich
SB-206553 hydrochloride hydrate, ≥98% (HPLC), solid