Merck
  • Intriguing hierarchical Co@NC microflowers in situ assembled by nanoneedles: Towards enhanced reduction of nitroaromatic compounds via interfacial synergistic catalysis.
Intriguing hierarchical Co@NC microflowers in situ assembled by nanoneedles: Towards enhanced reduction of nitroaromatic compounds via interfacial synergistic catalysis.
Journal of hazardous materials (2020-12-04)
Jiangyong Liu, Jinxing Li, Panming Jian, Ruiqi Jian
ABSTRACT
Developing highly efficient and cost-effective catalyst with tuned microstructure holds great promise in the reduction of nitroaromatic compounds under mild reaction conditions. Herein, we report a new Co@NC-MF catalyst with a fascinating hierarchical flower-like architecture in situ assembled from uniform Co@NC nanoneedles, which can function as a favorable platform for the efficient reduction of nitroaromatic compounds in the presence of NaBH4. In addition with the structural advantage, the characterization and experimental results demonstrate the enormous advantage of interfacial synergistic catalysis in enhancing the catalytic performance. The outside electron-rich N-doped carbon layer as Lewis basic sites and the inside Co nanoparticles are responsible for the adsorption of 4-nitrophenol (4-NP) and generation of active hydrogen species, respectively. This work contributes to the construction of well-integrated composites with well-balanced interface synergy to boost the catalytic performance in various heterogeneous reactions.
Sigma-Aldrich
2-NP, ≥98% (HPLC)