Merck
  • Resistance to PD1 blockade in the absence of metalloprotease-mediated LAG3 shedding.
Resistance to PD1 blockade in the absence of metalloprotease-mediated LAG3 shedding.
Science immunology (2020-07-19)
Lawrence P Andrews, Ashwin Somasundaram, Jessica M Moskovitz, Andrea L Szymczak-Workman, Chang Liu, Anthony R Cillo, Huang Lin, Daniel P Normolle, Kelly D Moynihan, Ichiro Taniuchi, Darrell J Irvine, John M Kirkwood, Evan J Lipson, Robert L Ferris, Tullia C Bruno, Creg J Workman, Dario A A Vignali
ABSTRACT
Mechanisms of resistance to cancer immunotherapy remain poorly understood. Lymphocyte activation gene-3 (LAG3) signaling is regulated by a disintegrin and metalloprotease domain-containing protein-10 (ADAM10)- and ADAM17-mediated cell surface shedding. Here, we show that mice expressing a metalloprotease-resistant, noncleavable LAG3 mutant (LAG3NC) are resistant to PD1 blockade and fail to mount an effective antitumor immune response. Expression of LAG3NC intrinsically perturbs CD4+ T conventional cells (Tconvs), limiting their capacity to provide CD8+ T cell help. Furthermore, the translational relevance for these observations is highlighted with an inverse correlation between high LAG3 and low ADAM10 expression on CD4+ Tconvs in the peripheral blood of patients with head and neck squamous cell carcinoma, which corresponded with poor prognosis. This correlation was also observed in a cohort of patients with skin cancers and was associated with increased disease progression after standard-of-care immunotherapy. These data suggest that subtle changes in LAG3 inhibitory receptor signaling can act as a resistance mechanism with a substantive effect on patient responsiveness to immunotherapy.
Sigma-Aldrich
Human LAG3 / Lymphocyte Activation Gene 3 Protein ELISA Kit, for serum, plasma, cell culture supernatants and urine
Pricing and availability is not currently available.