Merck
  • Determination and Comparison of Short-Chain Fatty Acids in Serum and Colon Content Samples: Alzheimer's Disease Rat as a Case Study.
Determination and Comparison of Short-Chain Fatty Acids in Serum and Colon Content Samples: Alzheimer's Disease Rat as a Case Study.
Molecules (Basel, Switzerland) (2020-12-10)
Lin-Xiu Guo, Yue Tong, Jue Wang, Guo Yin, Hou-Shuang Huang, Long Zeng, Ping Wang, Jun-Peng Li, Kai-Shun Bi, Tie-Jie Wang
ABSTRACT
Short-chain fatty acids (SCFAs) are the main microbial fermentation products from dietary fibers in the colon, and it has been speculated that they play a key role in keeping healthy in the whole-body. However, differences in SCFAs concentration in the serum and colon samples had attracted little attention. In this study, we have optimized the extract and analysis methods for the determination of ten SCFAs in both serum and colon content samples. Methanol and acetonitrile were chosen for extraction of SCFAs from serum and colon content samples, respectively. Biological samples were collected from Alzheimer's disease rats treated by extract of Schisandra chinensis (Turcz.) Baill (SC-extract) were taken as research objects. The results showed that, the relative peak intensities of SCFAs in the colon content from all groups were quite similar, and the trend was identical in the serum samples. Compared with the values in humans, the ratio of ten SCFAs in rat's colon was similar, while the percent of acetate in rat's serum was significantly higher. For therapy of Alzheimer's disease (AD), SC-extract decreased the concentration of butyrate, 3-Methyvalerate, and caproate in the serum samples towards the trend of normal rats. This study may help our understanding of how SCFAs are transported across colonic epithelium in healthy and diseased organisms.
Sigma-Aldrich
3-Nitrophenylhydrazine hydrochloride, 98%