Merck
  • Enhanced Thermal Sensitivity of TRPV3 in Keratinocytes Underlies Heat-Induced Pruritogen Release and Pruritus in Atopic Dermatitis.
Enhanced Thermal Sensitivity of TRPV3 in Keratinocytes Underlies Heat-Induced Pruritogen Release and Pruritus in Atopic Dermatitis.
The Journal of investigative dermatology (2020-04-06)
Seong Hoon Seo, Sohyun Kim, Song-Ee Kim, Seungsoo Chung, Sang Eun Lee
ABSTRACT
Itch in atopic dermatitis (AD) is aggravated under warm conditions. Transient receptor potential vanilloid (TRPV) 3, a member of the thermosensitive transient receptor potential channels, is activated by innocuous heat and is abundantly expressed in keratinocytes. The potential role of TRPV3 in itch is illustrated in TRPV3 channelopathies of humans and mice. However, the role of TRPV3 in heat-induced itch in AD and the underlying mechanisms are unclear. Here we showed that keratinocytes isolated from patients with AD exhibit enhanced expression and heat sensitivity with hyperactive channel function of TRPV3. Heat stimulus induced enhanced secretion of thymic stromal lymphopoietin, nerve growth factor, and prostaglandin E2 by keratinocytes from patients with AD through TRPV3 activation. TRPV3 agonists increased thymic stromal lymphopoietin, nerve growth factor, prostaglandin E2, and IL-33 production in human keratinocytes and induced scratching behavior upon intradermal injection in mice. TRPV3 was upregulated in the skin of MC903-induced AD mouse model. Heat stimulation to MC903-treated mice increased scratching behavior and produced higher levels of thymic stromal lymphopoietin, nerve growth factor, prostaglandin E2, and IL-33 from the epidermis, which were attenuated by pharmacologic inhibition of TRPV3. Moreover, neutralization of thymic stromal lymphopoietin reduced heat-evoked scratching in MC903-challenged mice. These results suggest that TRPV3 is a potential therapeutic target for heat-induced itch in AD.
Product Description
Supelco
2-Aminoethyl diphenylborinate, for TLC derivatization, ≥97.0%
Sigma-Aldrich
Monoclonal Anti-Nerve Growth Factor-β antibody produced in mouse, clone 25623, purified immunoglobulin, lyophilized powder