Merck
  • Organic Micropollutants in Wastewater Effluents and the Receiving Coastal Waters, Sediments, and Biota of Lyttelton Harbour (Te Whakaraupō), New Zealand.
Organic Micropollutants in Wastewater Effluents and the Receiving Coastal Waters, Sediments, and Biota of Lyttelton Harbour (Te Whakaraupō), New Zealand.
Archives of environmental contamination and toxicology (2020-11-01)
Philipp Emnet, Anjula Sachintha Mahaliyana, Grant Northcott, Sally Gaw
ABSTRACT
Coastal ecosystems are receiving environments for micropollutants due to high levels of associated anthropogenic activities. Effluent discharges from wastewater treatment plants are a significant source of micropollutants to coastal environments. Wastewater effluents, seawater, sediments, and green-lipped mussels (Perna canaliculus) in Lyttelton Harbour (Te Whakaraupō), Christchurch, New Zealand, were analysed for a suite of personal care products and steroid hormones during a 1-year period. In wastewater effluents, the concentration of methyl paraben (mParaben), ethyl paraben (eParaben), propyl paraben (pParaben), butyl paraben (bParaben), 4-t-octylphenol (OP), 4-methylbenzylidene camphor (4-MBC), benzophenone-3 (BP-3), benzophenone-1 (BP-1), triclosan, methyl triclosan (mTric), Bisphenol A (BPA), Estrone (E1), 17β-estradiol (E2), 17α-ethinyl estradiol (EE2), and Estriol (E3) ranged from < 0.6 to 429 ng L-1 and was dominated by OP, 4-MBC, BP-3, triclosan, BP-1, and BPA. In seawater, 4-MBC, BP-3, BPA, and E1 were the most frequently detected contaminants (< 0.2-9.4 ng L-1). Coastal sediment samples contained mParaben, OP, 4-MBC, BP-3, BP-1, BPA, OMC, and E1 (< 0.2-11 ng g-1 d.w.), and mParaben, OP, and BP-3 were found to bioaccumulate (3.8-21.3 ng g-1 d.w.) in green lipped mussels.
Sigma-Aldrich
2-Phenoxybenzoic acid, 98%