Merck
Double Porphyrin Cage Compounds.
European journal of organic chemistry (2021-01-01)
Kathleen Stout, Theo P J Peters, Mathijs F J Mabesoone, Fabian L L Visschers, Eline M Meijer, Joëlle-Rose Klop, Jeroen van den Berg, Paul B White, Alan E Rowan, Roeland J M Nolte, Johannes A A W Elemans
ABSTRACT
The synthesis and characterization of double porphyrin cage compounds are described. They consist of two porphyrins that are each attached to a diphenylglycoluril-based clip molecule via four ethyleneoxy spacers, and are linked together by a single alkyl chain using "click"-chemistry. Following a newly developed multistep synthesis procedure we report three of these double porphyrin cages, linked by spacers of different lengths, i.e. 3, 5, and 11 carbon atoms. The structures of the double porphyrin cages were fully characterized by NMR, which revealed that they consist of mixtures of two diastereoisomers. Their zinc derivatives are capable of forming sandwich-like complexes with the ditopic ligand 1,4-diazabicyclo[2,2,2]octane (dabco).
Sigma-Aldrich
Cesium triiodide, ≥99.9%
Pricing and availability is not currently available.