Merck
  • Electrochemically-deposited PANI on iron mesh-based metal-organic framework with enhanced visible-light response towards elimination of thiamphenicol and E. coli.
Electrochemically-deposited PANI on iron mesh-based metal-organic framework with enhanced visible-light response towards elimination of thiamphenicol and E. coli.
Environmental research (2020-08-21)
Jibin An, Yanlin Li, Wei Chen, Guoqiang Li, Jiahong He, Huixia Feng
ABSTRACT
Metal-organic frameworks (MOFs) are emerging class of porous materials that attracted tremendous attention as eco-friendly photocatalysts. However, poor charge separation in most MOFs largely thwarts their photocatalytic performance. In this work, Materials of Institut Lavoisier-100(Fe) (MIL-100 (Fe)) based on iron mesh was successfully fabricated by in situ growth. MIL-100(Fe) doped with polyaniline, namely MIL-100(Fe)/PANI, were then fabricated by galvanostatic deposition followed by annealing. Compared to pure MIL-100(Fe), MIL-100(Fe)/PANI composites exhibited excellent photocatalytic performances towards Thiamphenicol (TAP) degradation and Escherichia coli (E. Coli.) inactivation. The apparent rate constant, k, for TAP elimination of the MIL-100(Fe)/PANI composites with H2O2 is approximately 3 times as high as that of pure MIL-100(Fe). The electrochemical studies showed enhanced photocatalytic performances, which can be attributed to the electronic conductivity of PANI polymers. Quenching experiments, fluorescent tests and electron paramagnetic resonance (EPR) all suggested ⋅O2-, e-, ⋅OH and h+ as reactive oxidizing species (ROSs) involved in the photocatalytic process, where ⋅OH played the predominant ROSs. The transformation products of TAP were also isolated and characterized by high-resolution mass spectrometry, and transformation pathways of TAP under Vis/MIL-100(Fe)/PANI/H2O2 were tentatively clarified based on involved intermediates. Herein, MOFs conjugated conductive polymers nanocomposites look promising as efficient photocatalysts for organic pollutants degradation and bacteria inactivation. This work could offer novel strategies for the development of heterojunction composites with enhanced photocatalytic performances for better environmental remediation.
Sigma-Aldrich
BTC, ≥95% (HPLC)