Merck
  • The development of an immunoassay to measure immunoglobulin A in Asian elephant feces, saliva, urine and serum as a potential biomarker of well-being.
The development of an immunoassay to measure immunoglobulin A in Asian elephant feces, saliva, urine and serum as a potential biomarker of well-being.
Conservation physiology (2019-03-25)
Katie L Edwards, Pakkanut Bansiddhi, Steve Paris, Marie Galloway, Janine L Brown
ABSTRACT
Additional measures of well-being would be beneficial to the management of a variety of species in human care, including elephants. Immunoglobulin A (IgA) is an immune protein associated with pathogen defense, which has been demonstrated to decrease during times of stress, and increase in response to positive stimuli. This paper describes the development and validation of an enzyme immunoassay (EIA) for the quantification of Asian elephant (Elephas maximus) IgA in feces, saliva, urine, and serum. Samples were collected weekly from four females for 6 months to assess IgA and glucocorticoid (GC) concentrations, establish relationships between these two biomarkers, and determine variability in IgA within and between individuals, and across sample types. IgA was quantified in all four sample types, although urinary concentrations were low and sometimes undetectable in individual samples. Concentrations were highly variable within and between individuals, with fecal, salivary and serum IgA, and fecal, salivary and urinary GCs all differing significantly across individuals. Contrary to previous findings, IgA and GC were generally not correlated. Serum IgA was less variable within individuals, with the exception of one female that experienced a brief illness during the study. However, marked inter-individual differences were still apparent. When data from all individuals were combined, fecal IgA was significantly predicted by salivary and urinary IgA; however, this relationship did not hold when individuals were analyzed separately. Analysis of a fifth female that exhibited a more severe systemic illness demonstrated clear increases in fecal IgA and GC, suggesting these may also be useful health biomarkers. Further investigation is needed to determine what sample type is most reflective of biological state in elephants, and how IgA concentrations are associated with health and positive and negative welfare states. Based on observed variability, a longitudinal approach likely will be necessary to use IgA as a measure of well-being.
Sigma-Aldrich
IgA from human colostrum, reagent grade, buffered aqueous solution
Pricing and availability is not currently available.