Merck
  • Antioxidant activity toward fish oil triacylglycerols exerted by sphingoid bases isolated from butter serum with α-tocopherol.
Antioxidant activity toward fish oil triacylglycerols exerted by sphingoid bases isolated from butter serum with α-tocopherol.
Food chemistry (2020-07-30)
Ai Suzuki-Iwashima, Ai Iwasawa, Mayumi Kawai, Hiroaki Kubouchi, Ryuhei Ozaki, Kazuo Miyashita, Makoto Shiota
ABSTRACT
A mixture of sphingoid bases (SPGs) was prepared from butter serum, a by-product of anhydrous milk fat production. The mixture comprised seven types of SPGs with C16 to C19 alkyl chains. These milk SPGs inhibited the oxidation of fish oil triacylglycerol (TAG) more effectively than did a standard SPG (d18:1) with α-tocopherol. Reaction products were prepared from the combination of d18:0 or d18:1 with acrolein and propanal. Both sets of reaction products showed antioxidant activity toward fish oil TAG. Antioxidant activity of reaction products from d18:0 was stronger than that of reaction products from d18:1, suggesting that the molecule d18:0 may be a significant focus of the difference in antioxidant activity between milk SPGs and d18:1. To use SPGs as food additives in the future, an appropriate source of SPGs will be needed, and butter serum appears to have promise as a source of functional SPGs with strong antioxidant activity.
Sigma-Aldrich
2,4-Heptadienal, technical grade, 90%
Pricing and availability is not currently available.