Merck
  • Sulforaphane Elicits Protective Effects in Intestinal Ischemia Reperfusion Injury.
Sulforaphane Elicits Protective Effects in Intestinal Ischemia Reperfusion Injury.
International journal of molecular sciences (2020-07-28)
Zhiquan Chen, Annika Mohr, Barbara Heitplatz, Uwe Hansen, Andreas Pascher, Jens G Brockmann, Felix Becker
ABSTRACT
Intestinal ischemia reperfusion injury (IRI) is an inherent, unavoidable event of intestinal transplantation, contributing to allograft failure and rejection. The inflammatory state elicited by intestinal IRI is characterized by heightened leukocyte recruitment to the gut, which is amplified by a cross-talk with platelets at the endothelial border. Sulforaphane (SFN), a naturally occurring isothiocyanate, exhibits anti-inflammatory characteristics and has been shown to reduce platelet activation and block leukocyte adhesion. Thus, the aim of this study was to investigate protective effects and mechanism of action of SFN in a murine model of intestinal IRI. Intestinal IRI was induced by superior mesenteric artery occlusion for 30 min, followed by reperfusion for 2 h, 8 h or 24 h. To investigate cellular interactions, leukocytes were in vivo stained with rhodamine and platelets were harvested from donor animals and ex vivo stained. Mice (C57BL/6J) were divided into three groups: (1) control, (2) SFN treatment 24 h prior to reperfusion and (3) SFN treatment 24 h prior to platelet donation. Leukocyte and platelet recruitment was analyzed via intravital microscopy. Tissue was analyzed for morphological alterations in intestinal mucosa, barrier permeability, and leukocyte infiltration. Leukocyte rolling and adhesion was significantly reduced 2 h and 8 h after reperfusion. Mice receiving SFN treated platelets exhibited significantly decreased leukocyte and platelet recruitment. SFN showed protection for intestinal tissue with less damage observed in histopathological and ultrastructural evaluation. In summary, the data presented provide evidence for SFN as a potential therapeutic strategy against intestinal IRI.
Sigma-Aldrich
Germane, electronic grade, ≥99.997%
Sigma-Aldrich
o-Dianisidine dihydrochloride, ≥95%
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O