Merck
  • Oxidative stress, DNA stability and evoked inflammatory signaling in young celiac patients consuming a gluten-free diet.
Oxidative stress, DNA stability and evoked inflammatory signaling in young celiac patients consuming a gluten-free diet.
European journal of nutrition (2019-05-31)
Javier Diaz-Castro, Carlota Muriel-Neyra, Rafael Martin-Masot, Jorge Moreno-Fernandez, José Maldonado, Teresa Nestares
ABSTRACT
Celiac disease (CD) is a multifactorial, autoimmune, gluten-sensitive inflammatory disorder of the small intestine. Taking into account the pathogenesis of CD, a strict gluten-free diet (GFD) is the only treatment able to restore epithelium integrity and eliminate complications. The current study was designed to assess whether the use of a GFD is sufficient for maintaining a correct oxidative/antioxidant balance and ameliorating the evoked inflammatory signaling in young patients with CD. The study covered 80 children, aged between 7 and 18 years, attending the Gastroenterology Service of the Gastroenterology, Hepatology and Child Nutrition Service from the Virgen de las Nieves Hospital in Granada. Children with CD diagnosed were included in the celiac group who followed a strict GFD for 2 years (n = 40) and the control group (n = 40) included healthy children, with negative serological screening. Soluble superoxide dismutase 1 and 2, total antioxidant status, 8-hydroxy-2'-deoxyguanosine, cortisol, melatonin and inflammatory parameters in plasma, 15-F2t-isoprostanes in urine, and DNA breaks in peripheral blood lymphocytes were analysed. No differences were found in oxidative stress between CD patients and controls; however, IFN-γ, IL-1α, IP-10 and TNF-β were higher in the CD patients. VEGF was also higher than in the control group. The GFD in the CD patients is enough to reduce the oxidative stress; however, in the case of the inflammatory signaling, the initial exposure to gluten prior to stablish the GFD is strong enough to induce an inflammatory state which is maintained (even when consuming the GFD); meanwhile the increase in VEGF recorded in the CD group could be a compensatory mechanism to restore the damaged mucosa and duodenal villous atrophy, due to its role in endothelial activation and generation of new functional and stable vascular networks.
Product Description
Millipore
MILLIPLEX® Human Cytokine/Chemokine Magnetic Bead Panel - Premixed 29 Plex - Immunology Multiplex Assay, Simultaneously analyze multiple cytokine and chemokine biomarkers with Bead-Based Multiplex Assays using the Luminex technology, in human serum, plasma and cell culture samples.