Merck
  • Candida tropicalis is the most prevalent yeast species causing candidemia in Algeria: the urgent need for antifungal stewardship and infection control measures.
Candida tropicalis is the most prevalent yeast species causing candidemia in Algeria: the urgent need for antifungal stewardship and infection control measures.
Antimicrobial resistance and infection control (2020-04-09)
Youcef Megri, Amir Arastehfar, Teun Boekhout, Farnaz Daneshnia, Caroline Hörtnagl, Bettina Sartori, Ahmed Hafez, Weihua Pan, Cornelia Lass-Flörl, Boussad Hamrioui
ABSTRACT
Despite being associated with a high mortality and economic burden, data regarding candidemia are scant in Algeria. The aim of this study was to unveil the epidemiology of candidemia in Algeria, evaluate the antifungal susceptibility pattern of causative agents and understand the molecular mechanisms of antifungal resistance where applicable. Furthermore, by performing environmental screening and microsatellite typing we sought to identify the source of infection. We performed a retrospective epidemiological-based surveillance study and collected available blood yeast isolates recovered from the seven hospitals in Algiers. To identify the source of infection, we performed environmental screening from the hands of healthcare workers (HCWs) and high touch areas. Species identification was performed by API Auxa-Color and MALDI-TOF MS and ITS sequencing was performed for species not reliably identified by MALDI-TOF MS. Antifungal susceptibility testing followed CLSI M27-A3/S4 and included all blood and environmental yeast isolates. ERG11 sequencing was performed for azole-resistant Candida isolates. Microsatellite typing was performed for blood and environmental Candida species, where applicable. Candida tropicalis (19/66) was the main cause of candidemia in these seven hospitals, followed by Candida parapsilosis (18/66), Candida albicans (18/66), and Candida glabrata (7/66). The overall mortality rate was 68.6% (35/51) and was 81.2% for C. tropicalis-infected patients (13/16). Fluconazole was the main antifungal drug used (12/51); 41% of the patients (21/51) did not receive any systemic treatment. Candida parapsilosis was isolated mainly from the hands of HCWs (7/28), and various yeasts were collected from high-touch areas (11/47), including Naganishia albida, C. parapsilosis and C. glabrata. Typing data revealed interhospital transmission on two occasions for C. parapsilosis and C. glabrata, and the same clone of C. parapsilosis infected two patients within the same hospital. Resistance was only noted for C. tropicalis against azoles (6/19) and fluconazole-resistant C. tropicalis isolates (≥8 μg/ml) (6/19) contained a novel P56S (5/6) amino acid substitution and a previously reported one (V234F; 1/6) in Erg11p. Collectively, our data suggest an urgent need for antifungal stewardship and infection control strategies to improve the clinical outcome of Algerian patients with candidemia. The high prevalence of C. tropicalis joined by fluconazole-resistance may hamper the therapeutic efficacy of fluconazole, the frontline antifungal drug used in Algeria.
Sigma-Aldrich
Anidulafungin, ≥97% (HPLC)
Pricing and availability is not currently available.