Merck
  • A new perspective of lanthanide metal-organic frameworks: tailoring Dy-BTC nanospheres for rechargeable Li-O2 batteries.
A new perspective of lanthanide metal-organic frameworks: tailoring Dy-BTC nanospheres for rechargeable Li-O2 batteries.
Nanoscale (2020-04-22)
Dan Liu, Xinmin Zhang, Yan-Jie Wang, Shuyan Song, Lifeng Cui, Hongbo Fan, Xiaochang Qiao, Baizeng Fang
ABSTRACT
Nanoscaled lanthanide metal-organic frameworks (NLn-MOFs) have emerged as attractive nanomaterials for photofunctional applications. To enhance the inherent properties and endow NLn-MOF materials with desired electrochemical performance for rechargeable Li-O2 batteries, rational design and synthesis of NLn-MOFs with tailored morphologies for high O2 accessibility and rich open metal sites to bind O2 molecules is highly desired and remains a grand challenge. Herein, we prepare Dy-BTC nanospheres, which are explored for the first time as an O2 cathode in Li-O2 batteries. Interestingly, the specific capacity and electrochemical stability of the Dy-BTC nanosphere-based electrode outperform significantly those of the bulk crystalline Dy-BTC. A full discharge capacity of 7618 mA h g-1 at 50 mA g-1 has been achieved by the Dy-BTC nanospheres. Furthermore, the Dy-BTC nanospheres stably deliver a discharge capacity of 1000 mA h g-1 at 200 mA g-1 for 76 cycles, which is remarkably longer than that of the bulk crystalline Dy-BTC with a cycling life of 26 cycles.
Sigma-Aldrich
BTC, ≥95% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Dysprosium(III) nitrate hydrate, 99.9% trace metals basis
Pricing and availability is not currently available.