Merck
  • A bacterial riboswitch class senses xanthine and uric acid to regulate genes associated with purine oxidation.
A bacterial riboswitch class senses xanthine and uric acid to regulate genes associated with purine oxidation.
RNA (New York, N.Y.) (2020-04-30)
Diane Yu, Ronald R Breaker
ABSTRACT
Dozens of candidate orphan riboswitch classes have been discovered previously by using comparative sequence analysis algorithms to search bacterial genomic sequence databases. Each orphan is classified by the presence of distinct conserved nucleotide sequences and secondary structure features, and by its association with particular types of genes. One previously reported orphan riboswitch candidate is the "NMT1 motif," which forms a hairpin structure with an internal bulge that includes numerous highly conserved nucleotides. This motif associates with genes annotated to encode various dioxygenase enzymes, transporters, or proteins that have roles associated with thiamin or histidine metabolism. Biochemical evaluation of numerous ligand candidates revealed that NMT1 motif RNA constructs most tightly bind 8-azaxanthine, xanthine, and uric acid, whereas most other closely related compounds are strongly rejected. Genetic assays revealed that NMT1 motif RNAs function to turn off gene expression upon ligand binding, likely by regulating translation initiation. These results suggest that NMT1 motif RNAs function as aptamer domains for a riboswitch class that specifically responds to high concentrations of oxidized purines. Members of this "xanthine riboswitch" class appear to regulate genes predominantly related to purine transport and oxidation, thus avoiding the effects of overproduction of these common purine derivatives.