Merck
  • Transgenerational Inheritance of Betaine-Induced Epigenetic Alterations in Estrogen-Responsive IGF-2/IGFBP2 Genes in Rat Hippocampus.
Transgenerational Inheritance of Betaine-Induced Epigenetic Alterations in Estrogen-Responsive IGF-2/IGFBP2 Genes in Rat Hippocampus.
Molecular nutrition & food research (2020-02-06)
Yang Yang, Wenduo Jiang, Shu Yang, Fulei Qi, Ruqian Zhao
ABSTRACT
Betaine serves as a methyl donor for DNA methylation. Here, the effects of betaine on hippocampal expression of neurogenesis genes and their DNA methylation status across three generations are investigated. Pregnant rats (F0) are fed control and betaine-supplemented diets throughout gestation and lactation. Female F1 and F2 offspring at weaning, together with the F0 dams, are used in the study. Hippocampal expression of aromatase, estrogen receptor α, and estrogen-related receptor β is downregulated in F1, together with the estrogen-responsive insulin-like growth factor 2/insulin-like growth factor binding protein 2 (IGF-2/IGFBP2) genes. However, all these genes are upregulated in F2, which follows the same pattern of F0. In agreement with changes in mRNA expression, the imprinting control region (ICR) of IGF-2 gene is hypomethylated in F1 but hypermethylated in F2 and F0. In contrast, the promoter DNA methylation status of all the affected genes is hypermethylated in F1 but hypomethylated in F2 and F0. Methyl transfer enzymes, such as betaine homocysteine methyltransferase and DNA methyltransferase 1, follow the same pattern of transgenerational inheritance. These results indicate that betaine exerts a transgenerational effect on hippocampal expression of estrogen-responsive genes in rat offspring, which is associated with corresponding alterations in DNA methylation on ICR of IGF-2 gene and the promoter of affected genes.
Sigma-Aldrich
Betaine, ≥98% (perchloric acid titration)
Pricing and availability is not currently available.
Sigma-Aldrich
Betaine, ≥98%
Pricing and availability is not currently available.
Sigma-Aldrich
5-Methylcytidine, ≥99%
Pricing and availability is not currently available.