Merck
  • The Soybean Gene J Contributes to Salt Stress Tolerance by Up-Regulating Salt-Responsive Genes.
The Soybean Gene J Contributes to Salt Stress Tolerance by Up-Regulating Salt-Responsive Genes.
Frontiers in plant science (2020-04-08)
Qun Cheng, Zhuoran Gan, Yanping Wang, Sijia Lu, Zhihong Hou, Haiyang Li, Hongtao Xiang, Baohui Liu, Fanjiang Kong, Lidong Dong
ABSTRACT
Soybean [Glycine max (L.) Merr.] is an important crop for oil and protein resources worldwide, and its farming is impacted by increasing soil salinity levels. In Arabidopsis the gene EARLY FLOWERING 3 (ELF3), increased salt tolerance by suppressing salt stress response pathways. J is the ortholog of AtELF3 in soybean, and loss-of-function J-alleles greatly prolong soybean maturity and enhance grain yield. The exact role of J in abiotic stress response in soybean, however, remains unclear. In this study, we showed that J expression was induced by NaCl treatment and that the J protein was located in the nucleus. Compared to NIL-J, tolerance to NaCl was significantly lower in the NIL-j mutant. We also demonstrated that overexpression of J increased NaCl tolerance in transgenic soybean hairy roots. J positively regulated expression of downstream salt stress response genes, including GmWRKY12, GmWRKY27, GmWRKY54, GmNAC, and GmSIN1. Our study disclosed a mechanism in soybean for regulation of the salt stress response. Manipulation of these genes should facilitate improvements in salt tolerance in soybean.
Sigma-Aldrich
Spectrum Plant Total RNA Kit, sufficient for 10 purifications
Pricing and availability is not currently available.