Merck
Oxidative stability of spray dried matcha-tuna oil powders.
Food research international (Ottawa, Ont.) (2020-04-26)
Meng Shi, DanYang Ying, Mya Myintzu Hlaing, JianHui Ye, Luz Sanguansri, Mary Ann Augustin
ABSTRACT
Matcha-tuna oil and matcha-maltodextrin-tuna oil emulsions (25% oil, dry basis), formulated to have protein: carbohydrate ratios of 1:1.1, 1:2, 1:3 and 1:4, were spray dried. Confocal laser scanning microscopy showed effective emulsification of oil in all emulsions. All powders had low surface fat (2.9-4.2%). The addition of maltodextrin enhanced the bulk density and flowability of powders. Water sorption isotherms indicated that addition of maltodextrin increased water uptake of powders. The oxidative stability of the powders under accelerated conditions in an Oxipres® was highest for the matcha-tuna oil powder. Increasing amounts of added maltodextrin decreased oxidative stability. A comparison of levels of eicosapentaenoic acid (EPA) and docosahexaenoic acid (DHA) in neat oil and tuna oil powders over 12 weeks at 40 °C, demonstrated that % remaining EPA and DHA were higher for all spray dried powders compared to neat oil. There was a significant correlation (p < 0.01) between the amount of the loss of tea catechins and % remaining EPA and DHA after 12 weeks at 40 °C, suggesting that the catechins had a major role in protecting the tuna oil against oxidation. This study has demonstrated the potential of using a whole biomass (matcha) as the single encapsulant for protection and delivery of omega-3 oils.
Sigma-Aldrich
(−)-Catechin gallate, ≥98% (HPLC), from green tea
Pricing and availability is not currently available.